Assay Details
Gene Symbol: LOC107275966 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775401.2 | XP_015630887.1 | 1 - 2 | 862 | 77 |
Gene Symbol: LOC4332861 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777795.2 | XP_015633281.1 | 1 - 2 | 862 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
