Assay Details
| Gene Symbol: LOC107275966 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775401.2 | XP_015630887.1 | 3 - 4 | 1512 | 80 | |
| Gene Symbol: LOC4332861 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777795.2 | XP_015633281.1 | 3 - 4 | 1512 | 80 | |
More Information
Set Membership: |
|

LOC107275966
Genomic Map