Assay Details
Gene Symbol: LOC4333880 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015775645.2 | XP_015631131.1 | 1 - 2 | 229 | 146 | |
XM_015775647.2 | XP_015631133.1 | 1 - 2 | 229 | 146 | ||
GenBank mRNA | AK062077.1 | - | 1 - 2 | 162 | 146 | |
AK103065.1 | - | 1 - 2 | 159 | 146 | ||
EU268000.1 | - | 1 - 2 | 138 | 146 | ||
KC140113.1 | - | 1 - 2 | 199 | 146 |
Gene Symbol: LOC4339683 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK288455.1 | - | 2474 | 146 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015775645.2, XM_015775647.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
