Assay Details
| Gene Symbol: LOC4333880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775645.2 | XP_015631131.1 | 4 - 5 | 883 | 61 | |
| XM_015775646.2 | XP_015631132.1 | 4 - 5 | 833 | 61 | ||
| XM_015775647.2 | XP_015631133.1 | 4 - 5 | 883 | 61 | ||
| XM_015775648.2 | XP_015631134.1 | 4 - 5 | 827 | 61 | ||
| GenBank mRNA | AK062077.1 | - | 4 - 5 | 816 | 61 | |
| AK103065.1 | - | 4 - 5 | 813 | 61 | ||
| BT129313.1 | - | 3 - 4 | 333 | 61 | ||
| BT132195.1 | - | 2 - 3 | 159 | 61 | ||
| EU268000.1 | - | 4 - 5 | 792 | 61 | ||
| KC140113.1 | - | 4 - 5 | 853 | 61 | ||
| Gene Symbol: LOC4339683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK288455.1 | - | 1820 | 61 | ||
More Information
Set Membership: |
|

LOC4333880
Genomic Map