Assay Details
| Gene Symbol: LOC4326296 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766621.2 | XP_015622107.1 | 3 - 4 | 1165 | 78 | |
| XM_015766622.2 | XP_015622108.1 | 3 - 4 | 1165 | 78 | ||
| GenBank mRNA | AK058683.1 | - | 3 - 4 | 864 | 78 | |
| AK100613.1 | - | 3 - 4 | 1167 | 78 | ||
More Information
Set Membership: |
|

LOC4326296
Genomic Map