Assay Details
Gene Symbol: LOC4327139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766498.2 | XP_015621984.1 | 1 - 2 | 302 | 77 | |
GenBank mRNA | AK103697.1 | - | 1 - 2 | 202 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
