Assay Details
Gene Symbol: LOC4327139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766498.2 | XP_015621984.1 | 3 - 4 | 469 | 73 | |
GenBank mRNA | AK103697.1 | - | 3 - 4 | 369 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
