Assay Details
Gene Symbol: LOC4327139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766498.2 | XP_015621984.1 | 7 - 8 | 995 | 80 | |
GenBank mRNA | AK103697.1 | - | 7 - 8 | 895 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
