Assay Details
Gene Symbol: LOC9268869 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015755933.2 | XP_015611419.1 | 3 - 4 | 685 | 78 | |
GenBank mRNA | AK243305.1 | - | 3 - 4 | 686 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
