Assay Details
Gene Symbol: LOC9268869 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015755933.2 | XP_015611419.1 | 7 - 8 | 1188 | 120 | |
GenBank mRNA | AK243305.1 | - | 7 - 8 | 1189 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
