Assay Details
Gene Symbol: LOC4326190 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765494.1 | XP_015620980.1 | 2 - 3 | 608 | 79 | |
GenBank mRNA | AK122104.1 | - | 2 - 3 | 609 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
