Assay Details
| Gene Symbol: LOC4327922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762162.1 | XP_015617648.1 | 10 - 11 | 1778 | 73 | |
| XM_015762168.2 | XP_015617654.1 | 10 - 11 | 1763 | 73 | ||
| XM_015762176.1 | XP_015617662.1 | 10 - 11 | 1778 | 73 | ||
| XM_015762183.2 | XP_015617669.1 | 10 - 11 | 1759 | 73 | ||
| GenBank mRNA | AK061446.1 | - | 3 - 4 | 575 | 73 | |
| AK062187.1 | - | 1 - 2 | 274 | 73 | ||
| AK067665.1 | - | 10 - 11 | 1703 | 73 | ||
| AK099552.1 | - | 10 - 11 | 1601 | 73 | ||
More Information
Set Membership: |
|

LOC4327922
Genomic Map