Assay Details
| Gene Symbol: LOC4327922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762162.1 | XP_015617648.1 | 2 - 3 | 469 | 67 | |
| XM_015762168.2 | XP_015617654.1 | 2 - 3 | 454 | 67 | ||
| XM_015762176.1 | XP_015617662.1 | 2 - 3 | 469 | 67 | ||
| XM_015762183.2 | XP_015617669.1 | 2 - 3 | 450 | 67 | ||
| XM_015762193.1 | XP_015617679.1 | 2 - 3 | 469 | 67 | ||
| GenBank mRNA | AK065969.1 | - | 2 - 3 | 367 | 67 | |
| AK067665.1 | - | 2 - 3 | 369 | 67 | ||
| AK068876.1 | - | 2 - 3 | 411 | 67 | ||
| AK099552.1 | - | 2 - 3 | 292 | 67 | ||
More Information
Set Membership: |
|

LOC4327922
Genomic Map