Assay Details
| Gene Symbol: LOC4327922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762162.1 | XP_015617648.1 | 3 - 4 | 568 | 86 | |
| XM_015762168.2 | XP_015617654.1 | 3 - 4 | 553 | 86 | ||
| XM_015762176.1 | XP_015617662.1 | 3 - 4 | 568 | 86 | ||
| XM_015762183.2 | XP_015617669.1 | 3 - 4 | 549 | 86 | ||
| XM_015762193.1 | XP_015617679.1 | 3 - 4 | 568 | 86 | ||
| GenBank mRNA | AK065969.1 | - | 3 - 4 | 466 | 86 | |
| AK067665.1 | - | 3 - 4 | 468 | 86 | ||
| AK068876.1 | - | 3 - 4 | 510 | 86 | ||
| AK099552.1 | - | 3 - 4 | 391 | 86 | ||
More Information
Set Membership: |
|

LOC4327922
Genomic Map