Assay Details
Gene Symbol: LOC4327922 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015762162.1 | XP_015617648.1 | 6 - 7 | 1001 | 125 | |
XM_015762168.2 | XP_015617654.1 | 6 - 7 | 986 | 125 | ||
XM_015762176.1 | XP_015617662.1 | 6 - 7 | 1001 | 125 | ||
XM_015762183.2 | XP_015617669.1 | 6 - 7 | 982 | 125 | ||
XM_015762193.1 | XP_015617679.1 | 6 - 7 | 1001 | 125 | ||
GenBank mRNA | AK067665.1 | - | 6 - 7 | 901 | 125 | |
AK068876.1 | - | 6 - 7 | 943 | 125 | ||
AK099552.1 | - | 6 - 7 | 824 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
