Assay Details
| Gene Symbol: LOC4327922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762162.1 | XP_015617648.1 | 7 - 8 | 1183 | 102 | |
| XM_015762168.2 | XP_015617654.1 | 7 - 8 | 1168 | 102 | ||
| XM_015762176.1 | XP_015617662.1 | 7 - 8 | 1183 | 102 | ||
| XM_015762183.2 | XP_015617669.1 | 7 - 8 | 1164 | 102 | ||
| XM_015762193.1 | XP_015617679.1 | 7 - 8 | 1183 | 102 | ||
| GenBank mRNA | AK067665.1 | - | 7 - 8 | 1083 | 102 | |
| AK068876.1 | - | 7 - 8 | 1125 | 102 | ||
| AK099552.1 | - | 7 - 8 | 1006 | 102 | ||
More Information
Set Membership: |
|

LOC4327922
Genomic Map