Assay Details
| Gene Symbol: LOC4326889 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765648.2 | XP_015621134.1 | 5 - 6 | 948 | 69 | |
| GenBank mRNA | AK068983.1 | - | 5 - 6 | 802 | 69 | |
More Information
Set Membership: |
|

LOC4326889
Genomic Map