Assay Details
| Gene Symbol: LOC4327550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794238.2 | 3 - 4 | 774 | 72 | ||
| GenBank mRNA | AK072502.1 | - | 3 - 4 | 747 | 72 | |
| D30716.1 | - | 2 - 3 | 682 | 72 | ||
| X78143.1 | - | 3 - 4 | 726 | 72 | ||
More Information
Set Membership: |
|

LOC4327550
Genomic Map