Assay Details
| Gene Symbol: LOC112936036 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772229.2 | XP_015627715.1 | 7 - 8 | 1736 | 97 | |
| XM_015772230.2 | XP_015627716.1 | 7 - 8 | 1730 | 97 | ||
| GenBank mRNA | AK073907.1 | - | 7 - 8 | 1119 | 97 | |
More Information
Set Membership: |
|

LOC112936036
Genomic Map