Assay Details
| Gene Symbol: LOC4328577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770102.2 | XP_015625588.1 | 4 - 5 | 1003 | 82 | |
| XM_015770103.2 | XP_015625589.1 | 4 - 5 | 1001 | 82 | ||
| GenBank mRNA | AK069748.1 | - | 4 - 5 | 870 | 82 | |
More Information
Set Membership: |
|

LOC4328577
Genomic Map