Assay Details
Gene Symbol: LOC4329007 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770843.2 | XP_015626329.1 | 4 - 5 | 645 | 72 | |
GenBank mRNA | AK072052.1 | - | 4 - 5 | 625 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
