Assay Details
Gene Symbol: LOC107276277 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015767686.2 | XP_015623172.1 | 3 - 4 | 317 | 88 | |
GenBank mRNA | DQ201201.1 | - | 2 - 3 | 166 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
