Assay Details
Gene Symbol: LOC107276277 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015767686.2 | XP_015623172.1 | 5 - 6 | 484 | 79 | |
GenBank mRNA | DQ201201.1 | - | 4 - 5 | 333 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
