Assay Details
Gene Symbol: LOC107276277 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015767686.2 | XP_015623172.1 | 8 - 9 | 735 | 102 | |
GenBank mRNA | DQ201201.1 | - | 7 - 8 | 584 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
