Assay Details
| Gene Symbol: LOC4329753 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771196.2 | XP_015626682.1 | 3 - 4 | 395 | 62 | |
| GenBank mRNA | AK109600.1 | - | 3 - 4 | 401 | 62 | |
More Information
Set Membership: |
|

LOC4329753
Genomic Map