Assay Details
| Gene Symbol: LOC4329835 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772160.2 | XP_015627646.1 | 11 - 12 | 2839 | 85 | |
| GenBank mRNA | AK102601.1 | - | 11 - 12 | 2816 | 85 | |
More Information
Set Membership: |
|

LOC4329835
Genomic Map