Assay Details
| Gene Symbol: LOC4329835 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772160.2 | XP_015627646.1 | 4 - 5 | 2117 | 131 | |
| XM_015772161.2 | XP_015627647.1 | 4 - 5 | 2117 | 131 | ||
| GenBank mRNA | AK102601.1 | - | 4 - 5 | 2094 | 131 | |
More Information
Set Membership: |
|

LOC4329835
Genomic Map