Assay Details
Gene Symbol: LOC4329933 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771244.2 | XP_015626730.1 | 1 - 2 | 323 | 103 | |
GenBank mRNA | AK109586.1 | - | 1 - 2 | 326 | 103 | |
AK119319.1 | - | 1 - 2 | 313 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015771244.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
