Assay Details
Gene Symbol: LOC4329933 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771244.2 | XP_015626730.1 | 3 - 4 | 2807 | 154 | |
GenBank mRNA | AK119319.1 | - | 3 - 4 | 2797 | 154 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
