Assay Details
Gene Symbol: LOC4329933 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771244.2 | XP_015626730.1 | 4 - 5 | 2910 | 112 | |
GenBank mRNA | AK119319.1 | - | 4 - 5 | 2900 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
