Assay Details
| Gene Symbol: LOC4350004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK241764.1 | - | 1003 | 68 | ||
| AK287873.1 | - | 1003 | 68 | |||
| Gene Symbol: LOC4330472 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770199.2 | XP_015625685.1 | 2 - 3 | 404 | 68 | |
| GenBank mRNA | AK064357.1 | - | 2 - 3 | 376 | 68 | |
| AK102318.1 | - | 2 - 3 | 324 | 68 | ||
More Information
Set Membership: |
|

LOC4350004
Genomic Map