Assay Details
| Gene Symbol: LOC4330570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771947.2 | XP_015627433.1 | 4 - 5 | 473 | 146 | |
| XM_015771948.2 | XP_015627434.1 | 4 - 5 | 449 | 146 | ||
| GenBank mRNA | AK242364.1 | - | 4 - 5 | 470 | 146 | |
More Information
Set Membership: |
|

LOC4330570
Genomic Map