Assay Details
| Gene Symbol: LOC4330570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771947.2 | XP_015627433.1 | 5 - 6 | 583 | 103 | |
| XM_015771948.2 | XP_015627434.1 | 5 - 6 | 559 | 103 | ||
| GenBank mRNA | AK242364.1 | - | 5 - 6 | 580 | 103 | |
More Information
Set Membership: |
|

LOC4330570
Genomic Map