Assay Details
| Gene Symbol: LOC4330670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770380.1 | XP_015625866.1 | 12 - 13 | 2795 | 81 | |
| XM_015770381.1 | XP_015625867.1 | 12 - 13 | 2792 | 81 | ||
| XM_015770382.1 | XP_015625868.1 | 11 - 12 | 2774 | 81 | ||
| GenBank mRNA | AK059128.1 | - | 2 - 3 | 449 | 81 | |
| AK099845.1 | - | 11 - 12 | 2768 | 81 | ||
More Information
Set Membership: |
|

LOC4330670
Genomic Map