Assay Details
| Gene Symbol: LOC4330670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770380.1 | XP_015625866.1 | 4 - 5 | 950 | 70 | |
| XM_015770381.1 | XP_015625867.1 | 4 - 5 | 950 | 70 | ||
| XM_015770382.1 | XP_015625868.1 | 4 - 5 | 950 | 70 | ||
| GenBank mRNA | AK099845.1 | - | 4 - 5 | 944 | 70 | |
More Information
Set Membership: |
|

LOC4330670
Genomic Map