Assay Details
| Gene Symbol: LOC4330670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770380.1 | XP_015625866.1 | 8 - 9 | 1359 | 86 | |
| XM_015770381.1 | XP_015625867.1 | 8 - 9 | 1356 | 86 | ||
| XM_015770382.1 | XP_015625868.1 | 7 - 8 | 1338 | 86 | ||
| GenBank mRNA | AK099845.1 | - | 7 - 8 | 1332 | 86 | |
More Information
Set Membership: |
|

LOC4330670
Genomic Map