Assay Details
| Gene Symbol: LOC4330670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770380.1 | XP_015625866.1 | 10 - 11 | 2238 | 143 | |
| XM_015770381.1 | XP_015625867.1 | 10 - 11 | 2235 | 143 | ||
| XM_015770382.1 | XP_015625868.1 | 9 - 10 | 2217 | 143 | ||
| GenBank mRNA | AK099845.1 | - | 9 - 10 | 2211 | 143 | |
More Information
Set Membership: |
|

LOC4330670
Genomic Map