Assay Details
| Gene Symbol: LOC4331518 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777422.2 | XP_015632908.1 | 7 - 8 | 1399 | 99 | |
| GenBank mRNA | AK122069.1 | - | 7 - 8 | 1304 | 99 | |
More Information
Set Membership: |
|

LOC4331518
Genomic Map