Assay Details
Gene Symbol: LOC4331577 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777222.2 | XP_015632708.1 | 2 - 3 | 1040 | 97 | |
GenBank mRNA | AK068968.1 | - | 2 - 3 | 1025 | 97 | |
AK100092.1 | - | 2 - 3 | 1025 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
