Assay Details
Gene Symbol: LOC4331790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772902.2 | XP_015628388.1 | 6 - 7 | 1272 | 77 | |
XM_015772903.2 | XP_015628389.1 | 4 - 5 | 684 | 77 | ||
XM_015772904.2 | XP_015628390.1 | 4 - 5 | 677 | 77 | ||
GenBank mRNA | AK067994.1 | - | 6 - 7 | 1212 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
