Assay Details
Gene Symbol: LOC4331790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772902.2 | XP_015628388.1 | 7 - 8 | 1492 | 103 | |
XM_015772903.2 | XP_015628389.1 | 5 - 6 | 904 | 103 | ||
XM_015772904.2 | XP_015628390.1 | 5 - 6 | 897 | 103 | ||
GenBank mRNA | AK067994.1 | - | 7 - 8 | 1432 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
