Assay Details
Gene Symbol: LOC4331837 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777673.2 | XP_015633159.1 | 1 - 2 | 576 | 64 | |
GenBank mRNA | AK067807.1 | - | 1 - 2 | 576 | 64 | |
AK101197.1 | - | 1 - 2 | 579 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
