Assay Details
| Gene Symbol: LOC4331837 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777673.2 | XP_015633159.1 | 2 - 3 | 682 | 104 | |
| GenBank mRNA | AK067807.1 | - | 2 - 3 | 682 | 104 | |
| AK101197.1 | - | 2 - 3 | 685 | 104 | ||
More Information
Set Membership: |
|

LOC4331837
Genomic Map