Assay Details
Gene Symbol: LOC4331837 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777673.2 | XP_015633159.1 | 3 - 4 | 2550 | 82 | |
GenBank mRNA | AK067807.1 | - | 3 - 4 | 2550 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
