Assay Details
Gene Symbol: LOC4331998 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777585.2 | XP_015633071.1 | 2 - 3 | 525 | 64 | |
GenBank mRNA | AK058618.1 | - | 2 - 3 | 480 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
