Assay Details
| Gene Symbol: LOC4332041 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772537.2 | XP_015628023.1 | 5 - 6 | 607 | 65 | |
| GenBank mRNA | AK119546.1 | - | 5 - 6 | 525 | 65 | |
| AY129294.1 | - | 5 - 6 | 607 | 65 | ||
More Information
Set Membership: |
|

LOC4332041
Genomic Map