Assay Details
| Gene Symbol: LOC4332041 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772537.2 | XP_015628023.1 | 8 - 9 | 1064 | 57 | |
| GenBank mRNA | AK119546.1 | - | 8 - 9 | 982 | 57 | |
| AY129294.1 | - | 8 - 9 | 1064 | 57 | ||
More Information
Set Membership: |
|

LOC4332041
Genomic Map