Assay Details
| Gene Symbol: LOC4332950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777494.2 | XP_015632980.1 | 4 - 5 | 1482 | 61 | |
| XM_015777495.2 | XP_015632981.1 | 4 - 5 | 1473 | 61 | ||
| GenBank mRNA | AK110542.1 | - | 2 - 3 | 915 | 61 | |
| AK120709.1 | - | 4 - 5 | 1316 | 61 | ||
More Information
Set Membership: |
|

LOC4332950
Genomic Map