Assay Details
| Gene Symbol: LOC4333818 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777283.2 | XP_015632769.1 | 3 - 4 | 810 | 70 | |
| GenBank mRNA | AB099850.1 | - | 3 - 4 | 789 | 70 | |
| AK103565.1 | - | 3 - 4 | 797 | 70 | ||
| EU032588.1 | - | 4 - 5 | 735 | 70 | ||
More Information
Set Membership: |
|

LOC4333818
Genomic Map