Assay Details
Gene Symbol: LOC4333847 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772925.2 | XP_015628411.1 | 3 - 4 | 913 | 134 | |
GenBank mRNA | AK073877.1 | - | 3 - 4 | 918 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
